EMAIL THIS PAGE TO A FRIEND
Tierarztliche Praxis
[Anesthesia of horses with xylazine and ketamine. 2. Anesthesia in adult horses].
PMID 2183391
Abstract
Xylazine and Ketamine were used for anaesthesia in 250 adult horses. This combination is useful both for induction and maintenance of anaesthesia. The induction with Xylazine and Ketamine has been successful not only with foals but also with high risk patients such as horses with colic. During the maintenance of anaesthesia Xylazine/Ketamine can be used with injectable anaesthetics as well as with volatile anaesthetics at the same time. A form of injection anaesthesia for short-time surgical procedures like castration and wound repair is presented. The maintenance of a Thiobarbiturate/Guaifenesin anaesthesia with Xylazine/Ketamine seems to be suitable for field conditions.