EMAIL THIS PAGE TO A FRIEND
Biological procedures online
Media composition influences yeast one- and two-hybrid results.
PMID 21843345
Abstract
Although yeast two-hybrid experiments are commonly used to identify protein interactions, the frequent occurrence of false negatives and false positives hampers data interpretation. Using both yeast one-hybrid and two-hybrid experiments, we have identified potential sources of these problems: the media preparation protocol and the source of the yeast nitrogen base may not only impact signal range but also effect whether a result appears positive or negative. While altering media preparation may optimize signal differences for individual experiments, media preparation must be reported in detail to replicate studies and accurately compare results from different experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0626 Yeast Nitrogen Base Without Amino Acids, Yeast classification medium