EMAIL THIS PAGE TO A FRIEND
BMC musculoskeletal disorders
Calcitonin substitution in calcitonin deficiency reduces particle-induced osteolysis.
PMID 21843355
Abstract
Periprosthetic osteolysis is a major cause of aseptic loosening in joint arthroplasty. This study investigates the impact of CT (calcitonin) deficiency and CT substitution under in-vivo circumstances on particle-induced osteolysis in Calca -/- mice. We used the murine calvarial osteolysis model based on ultra-high molecular weight polyethylene (UHMWPE) particles in 10 C57BL/6J wild-type (WT) mice and twenty Calca -/- mice. The mice were divided into six groups: WT without UHMWPE particles (Group 1), WT with UHMWPE particles (Group 2), Calca -/- mice without UHMWPE particles (Group 3), Calca -/- mice with UHMWPE particles (Group 4), Calca -/- mice without UHMWPE particles and calcitonin substitution (Group 5), and Calca -/- mice with UHMWPE particle implantation and calcitonin substitution (Group 6). Analytes were extracted from serum and urine. Bone resorption was measured by bone histomorphometry. The number of osteoclasts was determined by counting the tartrate-resistant acid phosphatase (TRACP) + cells. Bone resorption was significantly increased in Calca -/- mice compared with their corresponding WT. The eroded surface in Calca -/- mice with particle implantation was reduced by 20.6% after CT substitution. Osteoclast numbers were significantly increased in Calca -/- mice after particle implantation. Serum OPG (osteoprotegerin) increased significantly after CT substitution. As anticipated, Calca -/- mice show extensive osteolysis compared with wild-type mice, and CT substitution reduces particle-induced osteolysis.