EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis and evaluation of an N-acetylglucosamine biosynthesis inhibitor.
PMID 21843880
Abstract
The structural rationale, synthesis and evaluation of an inhibitor designed to block glucosamine synthesis by competitively inhibiting the action of glutamine: fructose-6-phosphate amidotransferase and subsequently reducing the transformation of any glucosamine-6-phosphate formed to UDP-N-acetylglucosamine are described. The inhibitor 2-acetamido-2,6-dideoxy-6-sulfo-D-glucose (D-glucosamine-6-sulfonate) is an analog of glucosamine-6-phosphate in which the phosphate group in the latter is replaced with a sulfonic acid group. The inhibitor is designed to function by three different modes which together reduce UDP-N-acetylglucosamine synthesis. This reduction was confirmed by evaluating the effect of the inhibitor on bacterial cell-wall synthesis and by demonstrating that it inhibits acetylation of glucosamine-6-phosphate competitively and by acting as a surrogate substrate. Inhibition of glucosamine production or suitably activated glucosamine in bacteria leads to disruption of the peptidoglycan structure, which results in softening, bulging, deformation, fragility and lysis of the cells. These modifications were documented by scanning electron microscopy for bacteria treated with the inhibitor. They were observed for inhibitor concentrations in the 20 mg/mL range for Escherichia coli and Bacillus subtilis and the 5 mg/mL range for Rhizobium trifolii.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5509
D-Glucosamine 6-phosphate, ≥98%
C6H14NO8P
G4878
D-Glucosamine 6-phosphate sodium salt, ≥98%
C6H14NO8P