EMAIL THIS PAGE TO A FRIEND
International journal of environmental research and public health
Hypothalamic glial-to-neuronal signaling during puberty: influence of alcohol.
PMID 21845163
Abstract
Mammalian puberty requires complex interactions between glial and neuronal regulatory systems within the hypothalamus that results in the timely increase in the secretion of luteinizing hormone releasing hormone (LHRH). Assessing the molecules required for the development of coordinated communication networks between glia and LHRH neuron terminals in the basal hypothalamus, as well as identifying substances capable of affecting cell-cell communication are important. One such pathway involves growth factors of the epidermal growth factor (EGF) family that bind to specific erbB receptors. Activation of this receptor results in the release of prostaglandin-E(2) (PGE(2)) from adjacent glial cells, which then acts on the nearby LHRH nerve terminals to elicit release of the peptide. Another pathway involves novel genes which synthesize adhesion/signaling proteins responsible for the structural integrity of bi-directional glial-neuronal communication. In this review, we will discuss the influence of these glial-neuronal communication pathways on the prepubertal LHRH secretory system, and furthermore, discuss the actions and interactions of alcohol on these two signaling processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8008
Luteinizing hormone releasing hormone, ≥97% (HPLC)
C55H74N16O14