EMAIL THIS PAGE TO A FRIEND
Journal of Medicinal Chemistry
Halogenated 2'-benzoylpyridine thiosemicarbazone (XBpT) chelators with potent and selective anti-neoplastic activity: relationship to intracellular redox activity.
PMID 21846118