EMAIL THIS PAGE TO A FRIEND
Organic letters
Cascade reaction of isatins with heterocyclic ketene aminals: synthesis of imidazopyrroloquinoline derivatives.
PMID 21848302
Abstract
A concise and efficient route for the synthesis of highly substituted imidazopyrroloquinoline derivatives by simply refluxing a reaction mixture of different types of isatins and heterocyclic ketene aminals (HKAs) by acetic acid was developed. This method is suitable for combinatorial and parallel syntheses in drug discovery; consequently, a library of highly substituted imidazopyrroloquinoline derivatives was rapidly constructed using the present protocol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114618
Isatin, 97%
C8H5NO2