EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
2,2-Bis(bromomethyl)-1,3-propanediol (technical grade).
PMID 21850113
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
301930
2,2-Bis(bromomethyl)-1,3-propanediol, 98%
C5H10Br2O2