EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Bis(chloromethyl) ether and technical-grade chloromethyl methyl ether.
PMID 21850114
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100331
Chloromethyl methyl ether, technical grade
C2H5ClO