EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
p-Cresidine.
PMID 21850134
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46111
2-Methoxy-5-methylaniline, analytical standard
C8H11NO
103284
2-Methoxy-5-methylaniline, 99%
C8H11NO