EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Cupferron.
PMID 21850135
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
675636
Cupferron, 97%, reagent grade
C6H9N3O2