EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Danthron.
PMID 21850139
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D108103
1,8-Dihydroxyanthraquinone, 96%
C14H8O4