EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
2,3-Dibromo-1-propanol.
PMID 21850143
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D43050
2,3-Dibromo-1-propanol, 98%
C3H6Br2O