EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
4-Chloro-o-phenylenediamine.
PMID 21850148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108871
4-Chloro-o-phenylenediamine, 97%
C6H7ClN2