EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Inhibitors of ergosterol biosynthesis as crop protection agents.
PMID 2185090
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1100000
Ergosterol, European Pharmacopoeia (EP) Reference Standard
C28H44O