EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Design of chemical shift-switching 19F magnetic resonance imaging probe for specific detection of human monoamine oxidase A.
PMID 21851116
Abstract
Monoamine oxidase (MAO) A is a flavoenzyme that catalyzes the oxidation of biologically important monoamines and is thought to be associated with psychiatric disorders. Here, we report a strategy for rationally designing a (19)F magnetic resonance imaging probe for the specific detection of human MAO-A (hMAO-A) activity. Our designed (19)F probe was oxidized expeditiously by hMAO-A to produce 2-fluoro-4-nitrophenol via a spontaneous β-elimination mechanism. Concomitant with the structural change of the probe to the product, the (19)F chemical shift changed by 4.2 ppm, which was enough to visualize the probe and enzymatic product separately. Importantly, our probe achieved excellent discrimination of hMAO-A from its isoform hMAO-B.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
345067
2-Fluoro-4-nitrophenol, 99%
C6H4FNO3