EMAIL THIS PAGE TO A FRIEND
Basic & clinical pharmacology & toxicology
A transient inhibition and permanent lack of catechol-O-methyltransferase have minor effects on feeding pattern of female rodents.
PMID 21851556
Abstract
Abnormal feeding behaviours have long been linked to disruptions in brain dopaminergic activity. Dopamine is metabolized, amongst others, by catechol-O-methyltransferase (COMT). Normally, COMT only plays a subordinate role in dopamine metabolism. However, changes in COMT activity, especially in the prefrontal cortex, become more important during events that evoke dopamine release. The current study investigated the effect of acute COMT inhibition on feeding in Wistar rats and C57BL/6 mice using a selective, brain penetrating COMT inhibitor (OR-1139). Furthermore, the effect of a long-term lack of COMT on feeding behaviour was studied in COMT-deficient (COMT -/-) mice. Apart from following the gross feeding behaviour of fasted rats and mice, the first 4 hr of re-feeding were recorded with a video camera to allow a more detailed analysis of feeding microstructure. In the acute study, rats and mice received a single injection of OR-1139 (3, 10 or 30 mg/kg), just before the experiment. We found that rats and mice receiving OR-1139 had fewer very short meals but more long meals than the controls. Treated mice even ate more frequently than the controls, but other feeding parameters remained unchanged. Conversely, COMT -/- mice displayed an increased latency to initiate the first meal and spent less total time eating than wild-type mice. In conclusion, although decreased/lack of COMT activity did not robustly alter feeding behaviour of female rodents, we observed some alterations in the microstructure of feeding. However, these minor changes were highly dependent on the extent and fashion in which COMT was manipulated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R108
Ro 41-0960, solid
C13H8FNO5