EMAIL THIS PAGE TO A FRIEND
Irish veterinary journal
A study of 54 cases of left displacement of the abomasum: February to July 2005.
PMID 21851688
Abstract
Fifty-four cows with left displacement of the abomasum (LDA) submitted to the hospital facility at Riverview Veterinary Clinic from February to July 2005 were treated by right flank laparotomy and omentopexy. Five cows died (a survival rate 90.7%) and one cow (1.8%) was culled due to recurrence of the LDA post-operatively. Forty-one cows (76%) returned to good production post-operatively. Thirty-nine cows (72%) were pregnant six months after corrective surgery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32422
Ceftiofur hydrochloride, VETRANAL, analytical standard
C19H17N5O7S3 · HCl