EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Dyes metabolized to 3,3'-dimethylbenzidine (3,3'-dimethylbenzidine dye class).
PMID 21852830
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8533
o-Tolidine, ≥97%, powder
C14H16N2
31659
o-Tolidine, analytical standard
C14H16N2