EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Glycidol.
PMID 21852841
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480819
(R)-(+)-Glycidol, 97%, optical purity ee: 98% (GLC)
C3H6O2
G5809
Glycidol, 96%
C3H6O2