EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Kupffer cells influence parenchymal invasion and phenotypic orientation, but not the proliferation, of liver progenitor cells in a murine model of liver injury.
PMID 21854752
Abstract
Activation of myofibroblasts (MF) and extracellular matrix (ECM) deposition predispose the expansion and differentiation of liver progenitor cells (LPC) during chronic liver injury. Because Kupffer cells (KC) are active modulators of tissue response and fibrosis, we analyzed their role in a model of LPC proliferation. A choline-deficient diet, supplemented by ethionine (CDE) was administrated to C57Bl/6J mice that were depleted of KC by repeated injections of clodronate (CLO) and compared to PBS-injected mice. On CDE, massive KC activation was observed in the PBS group, but this was blunted in CLO-treated mice. The depletion of KC did not influence LPC proliferation but reduced their invasive behavior. Instead of being found far into the parenchyma, as was found in the PBS group (mean distance from portal vein: 209 μm), LPC of CLO mice remained closer to the portal area (138 μm), forming aggregates and phenotypically resembling cells of biliary lineage. Notably, removal of KC was also associated with a significant decrease in amount of MF and ECM and in the expression of profibrotic factors. Thus, besides ECM and MF, KC are also a significant component of the microenvironmental changes preceding LPC expansion. Depletion of KC may limit the LPC parenchymal invasion through a deficiency in chemoattracting factors, reduced activation of MF, and/or a paucity of the ECM framework necessary for cell motility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1260
L-Ethionine, ≥99% (TLC)
C6H13NO2S