EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis of 3-heteroarylthioquinoline derivatives and their in vitro antituberculosis and cytotoxicity studies.
PMID 21855182
Abstract
A series of 3-heteroarylthioquinoline derivatives has been synthesized by the Friedlander annulation of 2-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]-1-aryl-1-ethanone/2-(1,3-benzothiazol-2-ylsulfanyl)-1-aryl-1-ethanone/1-aryl-2-[(2-phenyl-2H-1,2,3,4-tetraazol-5-yl)sulfanyl]-1-ethanone with 2-aminobenzophenone in good yields using YbCl(3) as the catalyst. These compounds have been screened for their in vitro activity against Mycobacterium tuberculosis H37Rv (MTB) and among the 21 compounds screened, 2-[2-(4-bromophenyl)-4-phenyl-3-quinolyl]sulfanyl-5-methyl-1,3,4-thiadiazole (5d) and 2-[2-(4-chlorophenyl)-4-phenyl-3-quinolyl]sulfanyl-5-methyl-1,3,4-thiadiazole (5c) were found to be the most active compounds with MIC of 3.2 and 3.5 μM respectively against MTB. The cytotoxic effects against mouse fibroblasts (NIH 3T3) in vitro were evaluated for 5c and 5d, which displayed no toxic effects (IC(50) > 1000 μM) against the mouse fibroblast cell line NIH 3T3.