EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Quantitative analysis of myo-inositol in urine, blood and nutritional supplements by high-performance liquid chromatography tandem mass spectrometry.
PMID 21856255
Abstract
Myo-inositol plays key physiological functions, necessitating development of methodology for quantification in biological matrices. Limitations of current mass spectrometry-based approaches include the need for a derivatisation step and/or sample clean-up. In addition, co-elution of glucose may cause ion suppression of myo-inositol signals, for example in blood or urine samples. We describe an HPLC-MS/MS method using a lead-form resin based column online to a triple quadrupole tandem mass spectrometer, which requires minimum sample preparation and no derivatisation. This method allows separation and selective detection of myo-inositol from other inositol stereoisomers. Importantly, inositol was also separated from hexose monosaccharides of the same molecular weight, including glucose, galactose, mannose and fructose. The inter- and intra-assay variability was determined for standard solutions and urine with inter-assay coefficient of variation (CV) of 1.1% and 3.5% respectively, while intra-assay CV was 2.3% and 3.6%. Urine and blood samples from normal individuals were analysed.