EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Inhibition of aldose reductase prevents endotoxin-induced inflammation by regulating the arachidonic acid pathway in murine macrophages.
PMID 21856412
Abstract
The bacterial endotoxin lipopolysaccharide (LPS) is known to induce release of arachidonic acid (AA) and its metabolic products, which play important roles in the inflammatory process. We have shown earlier that LPS-induced signals in macrophages are mediated by aldose reductase (AR). Here we have investigated the role of AR in LPS-induced release of AA metabolites and their modulation using a potent pharmacological inhibitor, fidarestat, and AR siRNA ablation in RAW264.7 macrophages and AR-knockout mouse peritoneal macrophages and heart tissue. Inhibition or genetic ablation of AR prevented the LPS-induced synthesis and release of AA metabolites such as PGE2, TXB, PGI2, and LTBs in macrophages. LPS-induced activation of cPLA2 was also prevented by AR inhibition. Similarly, AR inhibition also prevented the calcium ionophore A23187-induced cPLA2 and LTB4 in macrophages. Further, AR inhibition by fidarestat prevented the expression of AA-metabolizing enzymes such as COX-2 and LOX-5 in RAW264.7 cells and AR-knockout mouse-derived peritoneal macrophages. LPS-induced expression of AA-metabolizing enzymes and their catalyzed metabolic products was significantly lower in peritoneal macrophages and heart tissue from AR-knockout mice. LPS-induced activation of redox-sensitive signaling intermediates such as MAPKs, transcription factor NF-κB, and EGR-1, a transcriptional regulator of mPGES-1, which in collaboration with COX-2 leads to the production of PGE2, was also significantly prevented by AR inhibition. Taken together, our results indicate that AR mediates LPS-induced inflammation by regulating the AA-metabolic pathway and thus provide a novel role for AR inhibition in preventing inflammatory complications such as sepsis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5523
REV 5901
C22H25NO2