EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Mutations to metabolic enzymes in cancer herald a need to unify genetics and biochemistry.
PMID 21857150
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1252
DL-Isocitric acid trisodium salt hydrate, ≥93%
C6H5Na3O7 · xH2O