EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Insulin-induced phospho-oligosaccharide stimulates amino acid transport in isolated rat hepatocytes.
PMID 2185744
Abstract
The ability of the insulin-induced phospho-oligosaccharide to stimulate amino acid transport was studied in isolated rat hepatocytes. At low alpha-aminoisobutyric acid concentrations (0.1 mM), both 100 nM-insulin and 10 microM-phospho-oligosaccharide doubled amino acid uptake after 2 h of incubation. This stimulation was prevented by 0.1 mM-cycloheximide or 5 micrograms of actinomycin D/ml, indicating that the phospho-oligosaccharide, like insulin, was acting via the synthesis of a high-affinity transport component. The effects of the phospho-oligosaccharide and of insulin were blocked by Ins2P (2.5 mM), but not by myo-inositol, inositol hexaphosphoric acid or several monosaccharides such as mannose, glucosamine and galactose. Both the temporal effect on amino acid entry and the extent of stimulation of this process by the phospho-oligosaccharide indicate that this molecule mimics, and may mediate, some of the long-term actions of insulin. However, the effects of phospho-oligosaccharide and insulin were not exactly the same, since the effect of insulin, but not of the phospho-oligosaccharide, was additive with that of glucagon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
652369
1-Amino-1-cyclobutanecarboxylic acid, 97%
C5H9NO2