EMAIL THIS PAGE TO A FRIEND
Natural product research
Essential oil composition of the fruits of Periploca laevigata Aiton subsp. angustifolia (Labill.) Markgraf (Apocynaceae - Periplocoideae).
PMID 21859258
Abstract
The essential oil of the fruits of Periploca laevigata Aiton subsp. angustifolia (Labill.) Markgraf (Apocynaceae) from Lampedusa Island was obtained by hydrodistillation and its composition was analysed. The analyses allowed the identification and quantification of 64 volatile compounds belonging to different classes. The most abundant compounds were nonacosane, heptacosane, hentriacontane and δ-cadinene. Among the volatile compounds identified in the fruits of P. laevigata subsp. angustifolia, 31 are present in other taxa of Apocynaceae, 19 have antimicrobial activity and four are pheromones for the butterfly Danaus chrysippus. The possible ecological role of the volatile compounds found is briefly discussed.
Related Materials