EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Dimethylvinyl chloride.
PMID 21860474
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123358
1-Chloro-2-methylpropene, 98%
C4H7Cl