EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Nitroarenes (selected): 1-Nitropyrene.
PMID 21860496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR305
1-Nitropyrene, BCR® certified Reference Material
C16H9NO2
N22959
1-Nitropyrene, ≥95%
C16H9NO2