EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
Phosphonoacetate biosynthesis: in vitro detection of a novel NADP(+)-dependent phosphonoacetaldehyde-oxidizing activity in cell-extracts of the marine Roseovarius nubinhibens ISM.
PMID 21861368
Abstract
A novel phosphonoacetaldehyde-oxidizing activity was detected in cell-extracts of the marine bacterium Roseovarius nubinhibens ISM grown on 2-aminoethylphosphonic acid (2-AEP; ciliatine). Extracts also contained 2-AEP transaminase and phosphonoacetate hydrolase activities. These findings indicate the existence of a biological route from 2-AEP via phosphonoacetaldehyde for the production of phosphonoacetate, which has not previously been shown to be a natural product. The three enzymes appear to constitute a previously-unreported pathway for the mineralization of 2-AEP which is a potentially important source of phosphorus in the nutrient-stressed marine environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
268674
2-Aminoethylphosphonic acid, 99%
C2H8NO3P
284270
Phosphonoacetic acid, 98%
C2H5O5P