EMAIL THIS PAGE TO A FRIEND
Metabolism: clinical and experimental
Sources of hepatic glycogen synthesis following a milk-containing breakfast meal in healthy subjects.
PMID 21862086
Abstract
During feeding, dietary galactose is a potential source of hepatic glycogen synthesis; but its contribution has not been measured to date. In the presence of deuterated water ((2)H(2)O), uridine diphosphate (UDP)-glucose derived from galactose is not enriched, whereas the remainder derived from glucose-6-phosphate (G6P) is enriched in position 2 to the same level as body water, assuming complete G6P-fructose-6-phosphate (F6P) exchange. Hence, the difference between UDP-glucose position 2 and body water enrichments reflects the contribution of galactose to glycogen synthesis relative to all other sources. In study 1, G6P-F6P exchange in 6 healthy subjects was quantified by supplementing a milk-containing breakfast meal with 10 g of [U-(2)H(7)]glucose and quantifying the depletion of position 2 enrichment in urinary menthol glucuronide. In study 2, another 6 subjects ingested (2)H(2)O and acetaminophen followed by an identical breakfast meal with 10 g of [1-(13)C]glucose to resolve direct/indirect pathways and galactose contributions to glycogen synthesis. Metabolite enrichments were determined by (2)H and (13)C nuclear magnetic resonance. In study 1, G6P-F6P exchange approached completion; therefore, the difference between position 2 and body water enrichments in study 2 (0.20% ± 0.03% vs 0.27% ± 0.03%, P < .005) was attributed to galactose glycogenesis. Dietary galactose contributed 19% ± 3% to glycogen synthesis. Of the remainder, 58% ± 5% was derived from the direct pathway and 22% ± 4% via the indirect pathway. The contribution of galactose to hepatic glycogen synthesis was resolved from that of direct and indirect pathways using a combination of (2)H(2)O and [1-(13)C]glucose tracers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3627
D-Fructose 6-phosphate disodium salt hydrate, ≥98%, amorphous powder
C6H11Na2O9P · xH2O