EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Phenoxybenzamine hydrochloride.
PMID 21863081
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B019
Phenoxybenzamine hydrochloride, ≥97%, powder
C18H22ClNO · HCl