EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Polycyclic aromatic hydrocarbons: 15 Listings - benz[a]anthracene, benzo[b]fluoranthene, benzo[j]fluoranthene, benzo[k]fluoranthene, benzo[a]pyrene, dibenz[a,h]acridine, dibenz[a,j]acridine, dibenz[a,h]anthracene, 7H-dibenzo[c,g]carbazole, dibenzo[a,e]pyrene, dibenzo[a,h]pyrene, dibenzo[a,i]pyrene, dibenzo[a,l]pyrene, indeno[1,2,3-cd]pyrene, 5-methylchrysene.
PMID 21863085
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR266
7H-Dibenzo[c,g]carbazole, BCR® certified Reference Material
C20H13N
48490
Benzo[b]fluoranthene, analytical standard
C20H12
BCR047
Benzo[b]fluoranthene, BCR® certified Reference Material
C20H12
30958
Benzo[b]fluoranthene, certified reference material, TraceCERT®
C20H12
SCB-002
Benzo[b]fluoranthene, vial of 1 g, analytical standard
C20H12
275336
Benzo[b]fluoranthene, 98%
C20H12
36955
Benzo[b]fluoranthene solution, 100 μg/mL in cyclohexane, analytical standard
C20H12
48637
Benzo[b]fluoranthene solution, certified reference material, 200 μg/mL in methanol
C20H12
40072
Benzo[b]fluoranthene solution, certified reference material, 1000 μg/mL in acetone
C20H12
502332 Benzo[j]fluoranthene solution, certified reference material, 2000 μg/mL in dichloromethane
C20H12
392251
Benzo[k]fluoranthene, for fluorescence, ≥99%
C20H12
BCR048R
Benzo[k]fluoranthene, BCR® certified Reference Material
C20H12
03323
Benzo[k]fluoranthene, certified reference material, TraceCERT®
C20H12
48492
Benzo[k]fluoranthene, analytical standard
C20H12
48668
Benzo[k]fluoranthene solution, certified reference material, 200 μg/mL in methylene chloride
C20H12
40073
Benzo[k]fluoranthene solution, certified reference material, 1000 μg/mL in acetone
C20H12
49476-U
Benzo(k)fluoranthene solution, 100 μg/mL in methylene chloride, analytical standard
C20H12
BCR138
Dibenz[a,h]anthracene, BCR® certified Reference Material
C22H14
91861
Dibenz[a,h]anthracene, certified reference material, TraceCERT®
C22H14
SCD-003
Dibenz[a,h]anthracene, vial of 1 g, analytical standard
C22H14
48574
Dibenz[a,h]anthracene, analytical standard
C22H14
502340 Dibenz[a,h]pyrene solution, 500 μg/mL in methylene chloride: benzene (3:1), analytical standard
ERD-014
Dibenz(a,j)acridine, vial of 25 mg, analytical standard
C21H13N
BCR133
Dibenzo[a,e]pyrene, BCR® certified Reference Material
C24H14
BCR159
Dibenzo[a,h]pyrene, BCR® certified Reference Material
C24H14
SCD-152
Dibenzo[a,h]pyrene, vial of 10 mg, analytical standard
C24H14
36947
Indeno[1,2,3-c,d]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
C22H12
48669
Indeno[1,2,3-c,d]pyrene solution, certified reference material, 200 μg/mL in methanol
C22H12
94377
Indeno[1,2,3-cd]pyrene, certified reference material, TraceCERT®
C22H12
ERI-001
Indeno[1,2,3-cd]pyrene, vial of 25 mg, analytical standard
C22H12