EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
β‑Propiolactone.
PMID 21863089
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5648
β-Propiolactone, Grade II, ≥90%
C3H4O2