EMAIL THIS PAGE TO A FRIEND
Organic letters
An improved palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.
PMID 21863838
Abstract
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with greatly simplified conditions relative to our previous report.