EMAIL THIS PAGE TO A FRIEND
Toxicology
Prior or coinstantaneous oral exposure to environmental immunosuppressive agents aggravates mite allergen-induced atopic dermatitis-like immunoreaction in NC/Nga mice.
PMID 21864637
Abstract
Immunosuppressive environmental chemicals may increase the potency of allergens and thereby play a role in the development of allergic diseases such as allergic rhinitis, asthma and atopic dermatitis (AD). This study's primary objective was to examine the mechanisms behind the development of allergic diseases and immunosuppression induced by some environmental chemicals. We focused on the aggravation of AD by the organophosphorus pesticide O,O-diethyl-O-4-nitro-phenylthiophosphate (parathion) and the organochlorine pesticide 1,1,1-trichloro-2,2-bis(4-methoxyphenyl)ethane (methoxychlor), in NC/Nga mice sensitized with extract of Dermatophagoides farinae (Df). NC/Nga mice were exposed orally to parathion or methoxychlor prior or coinstantaneous with sensitization with Df. The mice were subsequently challenged with Df. One day after the last challenge with Df, we analyzed dermatitis severity and expression of genes in the ear auricle, immunoglobulin (Ig) E and IgG(2a) levels in serum, and in auricular lymph nodes, T- or B-cell numbers and cytokine production. Prior exposure to parathion or methoxychlor induced marked increases in the following: dermatitis severity and gene expression in the ear auricle, IgE and IgG(2a) levels in serum, expression of surface antigens on helper T-cell and IgE-positive B-cell, production of Th1 and Th2 cytokines, and production of IgE in auricular lymph-node cells. In contrast, coinstantaneous exposure to parathion or methoxychlor yielded, at most, small but significant decreases in all parameters. Our results indicate that atopic dermatitis can be aggravated by prior exposure to immunosuppressive environmental chemicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36161
Methoxychlor, PESTANAL®, analytical standard
C16H15Cl3O2