EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Histone deacetylase activators: N-acetylthioureas serve as highly potent and isozyme selective activators for human histone deacetylase-8 on a fluorescent substrate.
PMID 21865040
Abstract
We report, for the first time, that certain N-acetylthiourea derivatives serve as highly potent and isozyme selective activators for the recombinant form of human histone deacetylase-8 in the assay system containing Fluor-de-Lys as a fluorescent substrate. The experimental data reveals that such activating feature is manifested via decrease in the K(m) value of the enzyme's substrate and increase in the catalytic turnover rate of the enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A22858
Acetylthiourea, 99%
C3H6N2OS