EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Reverse fosmidomycin derivatives against the antimalarial drug target IspC (Dxr).
PMID 21866890
Abstract
Reverse hydroxamate-based inhibitors of IspC, a key enzyme of the non-mevalonate pathway of isoprenoid biosynthesis and a validated antimalarial target, were synthesized and biologically evaluated. The binding mode of one derivative in complex with EcIspC and a divalent metal ion was clarified by X-ray analysis. Pilot experiments have demonstrated in vivo potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8682
Fosmidomycin sodium salt hydrate, ≥95% (NMR)
C4H10NO5P · xNa+ · yH2O