EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
Metabolic detoxication pathways for sterigmatocystin in primary tracheal epithelial cells: structural identification of glutathione adducts.
PMID 21866893