EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
A general framework for inhibitor resistance in protein kinases.
PMID 21867912
Abstract
Protein kinases control virtually every aspect of normal and pathological cell physiology and are considered ideal targets for drug discovery. Most kinase inhibitors target the ATP binding site and interact with residue of a hinge loop connecting the small and large lobes of the kinase scaffold. Resistance to kinase inhibitors emerges during clinical treatment or as a result of in vitro selection approaches. Mutations conferring resistance to ATP site inhibitors often affect residues that line the ATP binding site and therefore contribute to selective inhibitor binding. Here, we show that mutations at two specific positions in the hinge loop, distinct from the previously characterized "gatekeeper," have general adverse effects on inhibitor sensitivity in six distantly related kinases, usually without consequences on kinase activity. Our results uncover a unifying mechanism of inhibitor resistance of protein kinases that might have widespread significance for drug target validation and clinical practice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I100
5-Iodotubericidin, ≥90%, solid
C11H13IN4O4
T0642
Tubercidin, from Streptomyces tubercidicus, ~95%
C11H14N4O4