EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
A novel murine model of severe pulmonary arterial hypertension.
PMID 21868504
Abstract
The complex pathologies associated with severe pulmonary arterial hypertension (PAH) in humans have been a challenge to reproduce in mice due to the subtle phenotype displayed to PAH stimuli. Here we aim to develop a novel murine model of PAH that recapitulates more of the pathologic processes, such as complex vascular remodeling and cardiac indices, that are not characteristic of alternative mouse models. Inhibition of vascular endothelial growth factor receptor (VEGFR) with SU5416 combined with 3 weeks of chronic hypoxia was investigated. Hemodynamics, cardiac function, histological assessment of pulmonary vasculature, and molecular pathway analysis gauged the extent of PAH pathology development. The combination of VEGFR inhibition with chronic hypoxia profoundly exacerbated all measures of PAH-like pathology when compared with hypoxia alone (> 45 mm Hg right ventricular pressure, > 0.35 right ventricular hypertrophy). The changes in pulmonary vascular remodeling in response to hypoxia were further enhanced on SU5416 treatment. Furthermore, hypoxia/SU5416 treatment steadily decreased cardiac output, indicating incipient heart failure. Molecular analysis showed a dysregulated transforming growth factor-β/bone morphogenetic protein/Smad axis in SU5416- and/or hypoxia-treated mice as well as augmented induction of IL-6 and Hif-1α levels. These changes were observed in accordance with up-regulation of Tph1 and Pdgfr gene transcripts as well as a rise in platelet-rich serotonin. Biomarker analysis in response to VEGFR inhibition and/or hypoxia revealed distinct signatures that correlate with cytokine profiles of patients with idiopathic PAH. These data describe a novel murine model of PAH, which displays many of the hallmarks of the human disease, thus opening new avenues of investigation to better understand PAH pathophysiology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8442
SU 5416, ≥98% (HPLC)
C15H14N2O