EMAIL THIS PAGE TO A FRIEND
Journal of genetics
Characterization of two freshwater silurid catfish using conventional and molecular cytogenetic techniques.
PMID 21869482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2659
Chromomycin A3 from Streptomyces griseus, ≥95% (HPLC)
C57H82O26