EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Site-specific fluorescent labeling of nascent proteins on the translating ribosome.
PMID 21870811
Abstract
As newly synthesized proteins emerge from the ribosome, they interact with a variety of cotranslational cellular machineries that facilitate their proper folding, maturation, and localization. These interactions are essential for proper function of the cell, and the ability to study these events is crucial to understanding cellular protein biogenesis. To this end, we have developed a highly efficient method to generate ribosome-nascent chain complexes (RNCs) site-specifically labeled with a fluorescent dye on the nascent polypeptide. The fluorescent RNC provides real-time, quantitative information on its cotranslational interaction with the signal recognition particle and will be a valuable tool in elucidating the role of the translating ribosome in numerous biochemical pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
792551 H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH, 98% (HPLC)
C13H13NO5