EMAIL THIS PAGE TO A FRIEND
Expert opinion on drug metabolism & toxicology
Pharmacokinetic evaluation of vadimezan (ASA404, 5,6-dimethylxanthenone-4-acetic acid, DMXAA).
PMID 21870897
Abstract
Understanding the pharmacokinetics (PK) and pharmacodynamics of a drug is important to optimizing its use. Vadimezan is a tumor vascular-disrupting agent that acutely disrupts blood flow within tumors and induces innate tumor immunity. It has enhanced the activity of anticancer treatments in preclinical models and early phase trials, although one Phase III trial result was negative and another is yet to be reported. Areas covered in this review are the preclinical and human PK and the inter-relationship among PK, toxicity and efficacy of vadimezan as a single agent and in combination with other therapies. These data are derived from a literature search on Medline and also from conference proceedings, abstracts and trial reports available up to June 2011. The disappointing results of one Phase III trial, despite the promising randomized Phase II trial data, highlight the challenges in translational research, especially in selecting the optimal development strategy. This paper discusses how different scheduling of vadimezan could significantly enhance the anticancer efficacy of this drug in combination with other therapies, especially those that do not require concurrent corticosteroid administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5817
DMXAA, ≥98% (HPLC), solid
C17H14O4