EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
A point mutation in the Medicago sativa GSA gene provides a novel, efficient, selectable marker for plant genetic engineering.
PMID 21875626
Abstract
Bacterial selectable marker genes (SMG) conferring antibiotic resistance are valuable tools in plant genetic engineering, but public concern and regulatory requirements have stimulated the development of alternative selection systems. We have previously demonstrated that a mutated Synechococcus elongatus HemL gene encoding glutamate 1-semialdehyde aminotransferase (GSA) is an efficient SMG in alfalfa. In fact, GSA is irreversibly inhibited by gabaculine (3-amino-2,3-dihydrobenzoic acid), but the mutated enzyme is gabaculine insensitive. With the aim to develop a plant derived SMG, we cloned and sequenced the Medicago sativa GSA cDNA and reproduced one of the two mutations associated with gabaculine resistance in Synechococcus, a transversion resulting in a methionine to isoleucine (M→I) substitution. This mutated gene was assessed as a SMG in tobacco and alfalfa Agrobacterium transformation, in comparison with the wild type gene. In tobacco, about 43% of the leaf explants produced green shoots, whereas in alfalfa 47% of the explants produced green embryos in the presence of 30 μM gabaculine when the M→I GSA was introduced. Escapes were absent in tobacco and only 6% in alfalfa. No effect on the plant phenotype was noticed. We propose this new SMG as a widely acceptable alternative to those currently used.