EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Single-pot derivatisation strategy for enhanced gliotoxin detection by HPLC and MALDI-ToF mass spectrometry.
PMID 21879298
Abstract
Gliotoxin is produced by non-ribosomal peptide synthesis and secreted from certain fungi, including Aspergillus fumigatus. It is an epipolythiodioxopiperazine that contains an intact disulphide bridge and is the focus of intense research as a consequence of its negative immunomodulatory properties. Gliotoxin detection is generally enabled by reversed-phase-high-performance liquid chromatography (RP-HPLC), with absorbance detection (220-280 nm), or liquid chromatography-mass spectrometry, yet detection is not readily achievable by matrix-assisted laser desorption ionisation-time-of-flight mass spectrometry (MALDI-ToF MS). We have developed a single-pot derivatisation strategy which uses sodium borohydride-mediated reduction of gliotoxin followed by immediate alkylation of exposed thiols by 5'-iodoacetamidofluorescein to yield a stable product, diacetamidofluorescein-gliotoxin (GT-(AF)(2)), of molecular mass 1103.931 Da ((M+H)+). This product is readily detectable by RP-HPLC and exhibits a 6.8-fold increase in molar absorptivity compared with gliotoxin, which results in a higher sensitivity of detection (40 ng; 125 pmoL). GT-(AF)(2) also fluoresces (excitation/emission, 492:518 nm). Unlike free gliotoxin, the product (>800 fmol) is detectable by MALDI-ToF MS. Sporidesmin A can also be detected by RP-HPLC and MALDI-ToF MS (>530 fmol) using this strategy. We also demonstrate that the strategy facilitates detection of gliotoxin (mean ± SD = 3.55 ± 0.07 μg 100 μL(-1); n = 2) produced by A. fumigatus, without the requirement for organic extraction of culture supernatants and associated solvent removal. GT-(AF)(2) is also detectable (150 ng; 460 pmol) by thin-layer chromatography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9893
Gliotoxin from Gliocladium fimbriatum
C13H14N2O4S2