EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Polymorph formation and nucleation mechanism of tolfenamic acid in solution: an investigation of pre-nucleation solute association.
PMID 21879384
Abstract
Crystallization from solution involves nucleation and growth; growth conditions greatly influence self-association behaviors of solute molecules in these steps, affecting crystal packing of organic molecules. We examined the role of pre-nucleation association to provide insights into the mutual influence between molecular conformation in solution and packing in the solid state. Crystallization experiments of tolfenamic acid were conducted in ethanol under different supersaturation conditions. UV spectroscopy was performed to study self-association of solute molecules in ethanol as a function of concentration. Intermolecular interaction energies of tolfenamic acid dimers were calculated with quantum mechanical methods. As supersaturation increased, growth of the most stable polymorph outpaced the metastable one, contradicting Ostwald's Rule of Stages. UV spectroscopy measurement suggests solute molecules exist as hydrogen-bonded dimers and more dimers form as total concentration increases. Hydrogen bonding in the most stable form is significantly stronger than that in the metastable form. With the fact that molecular conformation is different in the two polymorphs, as concentration increases, solute molecules rearrange their conformations to form stronger hydrogen-bonded dimers in solution, resulting in nucleation of the most stable form.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0535
Tolfenamic acid, NSAID
C14H12ClNO2
34073
Tolfenamic acid, VETRANAL®, analytical standard
C14H12ClNO2