EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient copper-catalyzed S-vinylation of thiols with vinyl halides.
PMID 21879727
Abstract
The synthesis of vinyl sulfides through the coupling reaction of thiols with vinyl iodides, bromides, and chlorides is described. The thiols can couple with aryl iodides in the presence of only 0.5 mol % Cu(2)O without the need for an ancillary ligand. In the presence of 5 mol % of Cu(2)O and 10 mol % 1,10-phenanthroline as the ligand, the more challenging alkyl vinyl bromides can also be coupled with thiols, giving the vinyl sulfides in good to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1902
Vinyl bromide, 98%
C2H3Br
434191
Vinyl bromide solution, 1.0 M in THF
C2H3Br