EMAIL THIS PAGE TO A FRIEND
The Plant journal : for cell and molecular biology
Dynamic evolution of herbivore-induced sesquiterpene biosynthesis in sorghum and related grass crops.
PMID 21880075
Abstract
Sorghum (Sorghum bicolor) plants damaged by insects emit a blend of volatiles, predominantly sesquiterpenes, that are implicated in attracting natural enemies of the attacking insects. To characterize sesquiterpene biosynthesis in sorghum, seven terpene synthase (TPS) genes, SbTPS1 through SbTPS7, were identified based on their evolutionary relatedness to known sesquiterpene synthase genes from maize and rice. While SbTPS6 and SbTPS7 encode truncated proteins, all other TPS genes were determined to encode functional sesquiterpene synthases. Both SbTPS1 and SbTPS2 produced the major products zingiberene, β-bisabolene and β-sesquiphellandrene, but with opposite ratios of zingiberene to β-sesquiphellandrene. SbTPS3 produced (E)-α-bergamotene and (E)-β-farnesene. SbTPS4 formed (E)-β-caryophyllene as the major product. SbTPS5 produced mostly (E)-α-bergamotene and (Z)-γ-bisabolene. Based on the genome sequences of sorghum, maize and rice and the sesquiterpene synthase genes they contain, collinearity analysis identified the orthologs of sorghum sesquiterpene synthase genes, except for SbTPS4, in maize and rice. Phylogenetic analysis implied that SbTPS1, SbTPS2 and SbTPS3, which exist as tandem repeats, evolved as a consequence of local gene duplication in a lineage-specific manner. Structural modeling and site-directed mutagenesis experiments revealed that three amino acids in the active site play critical roles in defining product specificity of SbTPS1, SbTPS2, SbTPS3 and their orthologs in maize and rice. The naturally occurring functional variations of sesquiterpene synthases within and between species suggest that multiple mechanisms, including lineage-specific gene duplication, subfunctionalization, neofunctionalization and pseudogenization of duplicated genes, have all played a role in the dynamic evolution of insect-induced sesquiterpene biosynthesis in grasses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73492
trans-β-Farnesene, analytical standard
C15H24