EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Nucleotides part LXXX: Synthesis of 3'-O fluorescence labeled thymidine derivatives and their 5'-O-triphosphates.
PMID 21888545
Abstract
A new labeling technique attaching a fluorescent pteridine derivative (3, 5) via a linker onto the 3'-OH group of 5'-O-dimethoxytritylthymidine (7) was developed to lead to the conjugates 8 and 11. After detritylation to give 9 and 12, the final conversion into the corresponding 5'-triphosphates (13, 14), which were isolated as sodium salts, was performed by known methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360139
5′-O-(4,4′-Dimethoxytrityl)thymidine, 98%
C31H32N2O7